missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thirty one elements solution, 10mg/L HNO3 matrix, Fisher chemical™
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
£193.00 - £1335.00
Specifications
Chemical Name or Material | Thirty-One Elements |
---|---|
Concentration | 10mg/L |
Packaging | Bottle |
Type | ICP/MS Calibration Multi-element Standard Solution |
Analyte or Component Names | Ag,Al,As,Ba,B,Cd,Ce,Co,Cr,Cu,Dy,Er,Gd,Ho,La,Li,Lu,Mn,Nd,Ni,P, Pb,Rb,Se,Sm,Sr,Tl,Tm,U,V,Zn |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
12906193
|
Fisher Chemical
MS007050 |
50 mL | Bottle | N/A | |||||
12916193
|
Fisher Chemical
MS 007 100 |
100 mL | Bottle |
£293.00
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12926193
|
Fisher Chemical
MS 007 250 |
250 mL | Bottle |
£1335.00
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Thirty one elements solution - Ag, Al, As, Ba, B, Cd, Ce, Co, Cr, Cu, Dy, Er, Gd, Ho, La, Li, Lu, Mn, Nd, Ni, P, Pb, Rb Se Sm Sr, Tl, Tm, U, V, Zn for ICP/MS 10mg/L HNO3 matrixSpecifications
Thirty-One Elements | |
Bottle | |
Ag,Al,As,Ba,B,Cd,Ce,Co,Cr,Cu,Dy,Er,Gd,Ho,La,Li,Lu,Mn,Nd,Ni,P, Pb,Rb,Se,Sm,Sr,Tl,Tm,U,V,Zn | |
Liquid |
10mg/L | |
ICP/MS Calibration Multi-element Standard Solution | |
2% HNO3 |
Safety and Handling
H412
P273;P501
missing translation for 'shelfLife' : 12 Months. (6 Months after opening)
Product Identifier
Hazard Category
Hazard Statement
Precautionary Statement
- Multi-element Standard Solution for ICP. 31 Components in HNO3 2% (MS 007)
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P273-Avoid release to the environment.
- P501-Dispose of contents/container to .
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title