missing translation for 'onlineSavingsMsg'
Learn More

Brand™ Blaubrand™ Thoma Counting Chambers

CE-marked according to IVD-Directive 98/79 CE. BRAND™ Blaubrand™ Thoma Counting Chambers are used only for counting thrombocytes and erythrocytes. Rulings correspond to the central large square of the Neubauer chamber. The mini squares have an area of 0.0025mm2 each. Large outer squares left intentionally uncompleted.

Brand:  Brand™ 718005

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 10054030

  • £57.80 / Each
In Stock
to see stock.
Add to basket

Description

Description

  • Rulings correspond to the central large square of the Neubauer chamber.
  • The mini squares have an area of 0.0025mm2 each.
  • Used for counting thrombocytes and erythrocytes.
  • CE-marked according to IVD-Directive 98/79 CE.
Specifications

Specifications

Counting Device
0.1mm
Counting thrombocytes and erythrocytes
1Each
IVD-Directive 98/79 CE
Without spring clips
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers