Learn More
SymLine™ Adapters
Dovetail your SymLine™ System to suit your exact needs. Here, you will find many useful ancillaries.
Brand: SymLine™ 106410
37.96 GBP valid until 2024-12-31
Use promo code "23290" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Already exists
SymLine™ offers a suitable adapter for every common thread, so that you will have no problems connecting different thread types.
- Many useful accessories such as manifolds, shut-off valves, cabinet feed-throughs or earthing cables allow you to configure your SymLine™ system to your needs
- All accessories are electrostatically dissipative and approved for use in laboratories with explosion hazards
Specifications
Thread Adapter, G1/2 in. (m) to 11 to 18 mm (ID) Tube | |
Black | |
HDPE-EC | |
Adapter |