Learn More
SymLine™ Adapters
Dovetail your SymLine™ System to suit your exact needs. Here, you will find many useful ancillaries.
Brand:  SymLine™ 107610
35.70 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10508710
  • £44.55 / Each
Estimated Shipment: 10-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Already exists
SymLine™ offers a suitable adapter for every common thread, so that you will have no problems connecting different thread types.
  • Many useful accessories such as manifolds, shut-off valves, cabinet feed-throughs or earthing cables allow you to configure your SymLine™ system to your needs
  • All accessories are electrostatically dissipative and approved for use in laboratories with explosion hazards
Specifications
Specifications
Adapter Exhaust Filter to Stainless Steel Coupling
Justrite Container
Adapter
Black
HDPE
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
SymLine™ Adapters > Adapters; Description: Adapter Exhaust Filter to Stainless Steel Coupling; Material: HDPE; Type: Adapter