Learn More
SymLine™ Adapters
Dovetail your SymLine™ System to suit your exact needs. Here, you will find many useful ancillaries.
Brand: SymLine™ 107622
75.98 GBP valid until 2024-12-31
Use promo code "23290" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Already exists
SymLine™ offers a suitable adapter for every common thread, so that you will have no problems connecting different thread types.
- Many useful accessories such as manifolds, shut-off valves, cabinet feed-throughs or earthing cables allow you to configure your SymLine™ system to your needs
- All accessories are electrostatically dissipative and approved for use in laboratories with explosion hazards
Specifications
Offset Adapter 90°, Long | |
Waste Container Safety Cap, Exhaust Filter | |
Adapter |
Black | |
HDPE |