missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SymLine™ Adapters
Dovetail your SymLine™ System to suit your exact needs. Here, you will find many useful ancillaries.
Brand: SymLine™ 107082
62.56 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Already exists
SymLine™ offers a suitable adapter for every common thread, so that you will have no problems connecting different thread types.
- Many useful accessories such as manifolds, shut-off valves, cabinet feed-throughs or earthing cables allow you to configure your SymLine™ system to your needs
- All accessories are electrostatically dissipative and approved for use in laboratories with explosion hazards
Specifications
Thread Adapter, S60/61 (f) to 2 in. Tri-sure (f) | |
Black | |
HDPE-EC | |
Adapter |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
SymLine™ Adapters > Adapters; Description: Thread Adapter, S60/61 (f) to 2 in. Tri-sure (f); Material: HDPE-EC; Type: Adapter
Spot an opportunity for improvement?Share a Content Correction