missing translation for 'onlineSavingsMsg'
Learn More

SymLine™ Adapters

Dovetail your SymLine™ System to suit your exact needs. Here, you will find many useful ancillaries.

Brand:  SymLine™ 121211

61.71 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 17327429

  • £76.65 / Each
Estimated Shipment: 11-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Already exists

SymLine™ offers a suitable adapter for every common thread, so that you will have no problems connecting different thread types.

  • Many useful accessories such as manifolds, shut-off valves, cabinet feed-throughs or earthing cables allow you to configure your SymLine™ system to your needs
  • All accessories are electrostatically dissipative and approved for use in laboratories with explosion hazards
Specifications

Specifications

Borehole Reduction Adapter
Black
HDPE-EC
54 to 75 mm
33 mm
Adapter
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
SymLine™ Adapters > Adapters; Description: Borehole Reduction Adapter; Material: HDPE-EC; Type: Adapter

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.