Learn More
Lenz Laborglasintrumente™ Three-Neck Round-Bottom Flasks
Three-neck round-bottom reaction flasks with two parallel side necks
Brand:  Lenz Laborglasintrumente™ 03304170
 View more versions of this product
Product Code. 10264191
  • £55.55 / Each
Estimated Shipment: 06-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Reaction flasks acc. to DIN 12392 and in addition to DIN
  • With two parallel side necks
  • With conical ground joints
  • With inscription label
  • Material: DURAN™
Specifications
Specifications
Center neck- NS 29/32, Side necks - NS 14/23
Borosilicate Glass (3.3)
Reaction flasks
1000 mL
1
131 mm