Learn More
Lenz Laborglasintrumente™ Three-Neck Round-Bottom Flasks
Three-neck round-bottom reaction flasks with two parallel side necks
£55.55 - £1090.00
Specifications
Material Borosilicate Glass (3.3)
No. per Pack 1
Product Type Reaction flasks
View More Specs
Products 5
Product Code Brand Capacity (Metric) Description Price Quantity & Availability  
Product Code Brand Capacity (Metric) Description Price Quantity & Availability  
10121761
View Documents
Lenz Laborglasintrumente™
03304237
100 mL Center neck- NS 29/32 , Side necks -NS 19/26
£60.60
Each
Estimated Shipment: 06-02-2025
to see stock.
Add to basket
10264191
View Documents
Lenz Laborglasintrumente™
03304170
1000 mL Center neck- NS 29/32, Side necks - NS 14/23
£55.55
Each
Estimated Shipment: 06-02-2025
to see stock.
Add to basket
10709031
View Documents
Lenz Laborglasintrumente™
03305476
3000 mL Center neck- NS 45/40 , Side necks -NS 29/32
£274.00
Each
Estimated Shipment: 20-01-2025
to see stock.
Add to basket
10718841
View Documents
Lenz Laborglasintrumente™
03304479
4000 mL Center neck- NS 29/32, Side necks - NS 29/32
£276.00
Each
Estimated Shipment: 24-01-2025
to see stock.
Add to basket
10798451
View Documents
Lenz Laborglasintrumente™
03305494
20,000 mL Center neck- NS 45/40 , Side necks -NS 29/32
£1090.00
Each
Estimated Shipment: 06-02-2025
to see stock.
Add to basket
Description
Description
  • Reaction flasks acc. to DIN 12392 and in addition to DIN
  • With two parallel side necks
  • With conical ground joints
  • With inscription label
  • Material: DURAN™
Specifications
Specifications
Borosilicate Glass (3.3)
1
Reaction flasks