missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lenz Laborglasintrumente™ Three-Neck Round-Bottom Flasks
Three-neck round-bottom reaction flasks with two parallel side necks
Brand: Lenz Laborglasintrumente™ 03305494
This item is not returnable.
View return policy
Description
- Reaction flasks acc. to DIN 12392 and in addition to DIN
- With two parallel side necks
- With conical ground joints
- With inscription label
- Material: DURAN™
Specifications
Center neck- NS 45/40 , Side necks -NS 29/32 | |
Borosilicate Glass (3.3) | |
Reaction flasks |
20,000 mL | |
1 | |
345 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction