Learn More
DWK Life Sciences Three Part Retaining Device, complete, for stopcock key threaded, SBW -J glass
To secure the tap plug in the ground DURAN™ single way stopcock.
Brand: DWK Life Sciences 286039009
Code : NEW
Additional Details : Weight : 0.20000kg
Description
The 3-part tap safety device holds and secures the threaded tap plug in the tap sleeve.
- 3-part, complete for SBW-J tap plug
- Greater safety through installation of the 3-part screw locking device
Specifications
Polypropylene | |
12.5 NS | |
For securing the threaded tap plug in the ground DURAN™ one-way tap |
No | |
100/Pk. |