Learn More
DWK Life Sciences Three Part Retaining Device, complete, for stopcock key threaded, SBW -J glass
To secure the tap plug in the ground DURAN™ single way stopcock.
Brand:  DWK Life Sciences 286039403
 View more versions of this product
Product Code. 10568031
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
The 3-part tap safety device holds and secures the threaded tap plug in the tap sleeve.
  • 3-part, complete for SBW-J tap plug
  • Greater safety through installation of the 3-part screw locking device
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Polypropylene
18.8 NS
For securing the threaded tap plug in the ground DURAN™ one-way tap
Not Autoclavable
100/Pk.