Learn More
Three-prong Clamps
Use this clamp for a variety of laboratory applications. Three-prong Clamps are suitable for articles from 1 to 70mm in diameter.
Brand:  S Murray SAMCO L001/01
Product Code. 12378149
  • £56.53 / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Diecast aluminum construction
  • Nickel-plated brass tommy screw
  • Rubber covered jaws
Specifications
Specifications
Clamp
1 to 75 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Three-prong Clamps > Adjustable Dia.: 1 to 75mm