missing translation for 'onlineSavingsMsg'
Learn More
Thulium(III) nitrate hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals Product Code.: 11376238

Thulium(III) nitrate hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Product Code. 11376238
10 g
Click to view available options
Quantity:
2 g
10 g
50 g
This item is not returnable. View return policy

Product Code. 11376238

Brand: Thermo Scientific Alfa Aesar 011203.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used as laboratory reagent, optical glasses, structural ceramics, catalysts, electrical components and photo-optical material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100641-15-4
Molecular Formula N3O9Tm
Molecular Weight (g/mol) 354.95
MDL Number MFCD00149875
InChI Key LLZBVBSJCNUKLL-UHFFFAOYSA-N
IUPAC Name thulium(3+) trinitrate
SMILES [Tm+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Physical Form Wet Crystalline Aggregates
Quantity 10 g
UN Number UN1477
Solubility Information Soluble in water.
Formula Weight 354.95 (Anhydrous)
Percent Purity 99.99%
Grade Reacton™
Assay (REO)
Chemical Name or Material Thulium(III) nitrate hydrate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number61200477 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number011203.04,011203.09
Lot NumberN17G002 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number011203.04,011203.09
Lot NumberN21A034 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number011203.04,011203.09
Lot NumberL11M13 Certificate TypeCertificate of Analysis Date29/11/2021 Catalog Number011203.04,011203.09
Lot NumberP12H022 Certificate TypeCertificate of Analysis Date29/11/2021 Catalog Number011203.04,011203.09

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thulium(III) nitrate hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.