missing translation for 'onlineSavingsMsg'
Learn More

Thulium(III) nitrate hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

POA - £628.13

Chemical Identifiers

CAS 100641-15-4
Molecular Formula N3O9Tm
Molecular Weight (g/mol) 354.95
MDL Number MFCD00149875
InChI Key LLZBVBSJCNUKLL-UHFFFAOYSA-N
IUPAC Name thulium(3+) trinitrate
SMILES [Tm+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
View More Specs

Productos 3
Código de producto Marca Quantity Precio Cantidad y disponibilidad  
Código de producto Marca Quantity Precio Cantidad y disponibilidad  
11386238
missing translation for 'viewDocuments'
Thermo Scientific Alfa Aesar
011203.04
2 g
£137.69
2g
Please to purchase this item. Need a web account? Register with us today!
11376238
missing translation for 'viewDocuments'
Thermo Scientific Alfa Aesar
011203.09
10 g
£628.13
10g
Please to purchase this item. Need a web account? Register with us today!
11345829
missing translation for 'viewDocuments'
Thermo Scientific Alfa Aesar
011203.18
50 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Descripción

Descripción

Used as laboratory reagent, optical glasses, structural ceramics, catalysts, electrical components and photo-optical material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Especificaciones

Identificadores de productos químicos

100641-15-4
354.95
LLZBVBSJCNUKLL-UHFFFAOYSA-N
[Tm+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
N3O9Tm
MFCD00149875
thulium(3+) trinitrate

Especificaciones

100641-15-4
N3O9Tm
UN1477
LLZBVBSJCNUKLL-UHFFFAOYSA-N
thulium(3+) trinitrate
354.95 (Anhydrous)
Reacton™
Thulium(III) nitrate hydrate
Wet Crystalline Aggregates
MFCD00149875
Soluble in water.
[Tm+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
354.95
99.99%
(REO)
Videos
Seguridad y manipulación

Seguridad y manipulación

GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H272-H315-H319-H335

missing translation for 'dotInformation' : Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: NITRATES, INORGANIC, N.O.S.

missing translation for 'einecsNumber' : 239-069-9

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documentos

Documentos

Certificados

RUO – Research Use Only