missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Thymolphthalexone Product Code.: 11313106

Thermo Scientific Chemicals Thymolphthalexone

Product Code. 11313106
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11313106

Brand: Thermo Scientific Chemicals L10962.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1913-93-5
Molecular Formula C38H44N2O12
Molecular Weight (g/mol) 720.77
MDL Number MFCD00001619
InChI Key AGXCADFSYJNMEG-UHFFFAOYNA-N
Synonym thymolphthalexone, thymolphthalein complexon, thymolphthalein complexone, thymolphthalexon, thymolphthalein-3,3'-bismethyliminodiacetic acid, 3-1-3-bis carboxymethyl amino methyl-4-hydroxy-5-isopropyl-2-methylphenyl-3-oxo-2-benzofuran-1-yl-6-hydroxy-5-isopropyl-2-methylphenyl methyl carboxymethyl amino acetic acid, n,n'-3-oxo-1 3h-isobenzofuranylidene bis 6-hydroxy-2-methyl-5-1-methylethyl-3,1-phenylene methylene bis n-carboxymethyl glycine, thymophthalexon, 3',3-bis n,n-di carboxymethyl aminomethyl thymolphthalein, 2,2',2,2'-3-oxo-1,3-dihydro-2-benzofuran-1,1-diyl bis 6-hydroxy-5-isopropyl-2-methyl-3,1-phenylene methylenenitrilo tetraacetic acidShow MoreShow Less
PubChem CID 112918
IUPAC Name 2-[[5-[1-[3-[[bis(carboxymethyl)amino]methyl]-4-hydroxy-2-methyl-5-propan-2-ylphenyl]-3-oxo-2-benzofuran-1-yl]-2-hydroxy-6-methyl-3-propan-2-ylphenyl]methyl-(carboxymethyl)amino]acetic acidShow MoreShow Less
SMILES CC(C)C1=CC(=C(C)C(C[NH+](CC(O)=O)CC([O-])=O)=C1O)C1(OC(=O)C2=CC=CC=C12)C1=CC(C(C)C)=C(O)C(C[NH+](CC(O)=O)CC([O-])=O)=C1C

Specifications

Quantity 5 g
Beilstein 1339087
Formula Weight 720.78
Chemical Name or Material Thymolphthalexone

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10251548 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberL10962.06,L10962.03
Lot Number10245178 Certificate TypeCertificate of Analysis Date09/01/2023 Catalog NumberL10962.06,L10962.03
Lot Number10151562 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL10962.06,L10962.03
Lot Number10184865 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL10962.06,L10962.03
Lot Number10235909 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL10962.06,L10962.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Thymolphthalexone >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.