Learn More
3M™ MonitorMark™ Time Temperature Indicator
Solves the challenge of monitoring thermal exposure of temperature-sensitive products during transportation and storage.
Brand:  3M™ MonitorMark™ 7000001890
Additional Details : Weight : 0.48500kg
 View more versions of this product
Product Code. 17268510
  • £1467.36 / Pack of 500
Estimated Shipment: 02-08-2024
to see stock.
Add to basket
Description
Description
  • Inexpensive solution for monitoring product exposure to temperatures above the specified limit over a 48 hour time period.
  • Self-adhesive backing for easy attachment to secondary packaging.
  • Easily-interpreted visual results.
  • Results indicate both exposure and relative time over which exposure occurred.
  • Removable film activation strip starts the monitoring process on demand for accuracy.
Specifications
Specifications
500 /Cs.
95 mm
50°F
3.74 in.
48 Hours
10°C