Learn More
3M™ MonitorMark™ Time Temperature Indicator
Solves the challenge of monitoring thermal exposure of temperature-sensitive products during transportation and storage.
£1467.36 - £2980.93
Product Code | Brand | Temperature (Metric) Operating | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Temperature (Metric) Operating | Price | Quantity & Availability | |||||
17268510
|
3M™ MonitorMark™
7000001890 |
10°C |
£1467.36
Pack of 500 |
Estimated Shipment: 02-08-2024
Log in to see stock. |
|||||
17278510
|
3M™ MonitorMark™
7000001929 |
22°C |
£2980.93
Pack of 500 |
Estimated Shipment: 02-08-2024
Log in to see stock. |
|||||
Description
- Inexpensive solution for monitoring product exposure to temperatures above the specified limit over a 48 hour time period.
- Self-adhesive backing for easy attachment to secondary packaging.
- Easily-interpreted visual results.
- Results indicate both exposure and relative time over which exposure occurred.
- Removable film activation strip starts the monitoring process on demand for accuracy.
Specifications
500 /Cs. | |
48 Hours |