missing translation for 'onlineSavingsMsg'
Learn More

Tin(Ii) Chloride, Honeywell

Reagent Grade, 98%

£39.80 - £444.64

Chemical Identifiers

CAS 7772-99-8
Molecular Formula Cl2Sn
Molecular Weight (g/mol) 189.61
MDL Number MFCD00011241
InChI Key AXZWODMDQAVCJE-UHFFFAOYSA-L
PubChem CID 24479
ChEBI CHEBI:78067
IUPAC Name dichlorotin
SMILES [Cl-].[Cl-].[Sn++]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15671830
View Documents
Honeywell Research Chemicals
208256-100G
100 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15691830
View Documents
Honeywell Research Chemicals
208256-500G
500 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15681830
View Documents
Honeywell Research Chemicals
208256-2KG
2 kg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • ≥95% for general laboratory use.
Specifications

Chemical Identifiers

7772-99-8
189.61
AXZWODMDQAVCJE-UHFFFAOYSA-L
CHEBI:78067
[Cl-].[Cl-].[Sn++]
Cl2Sn
MFCD00011241
24479
dichlorotin

Specifications

7772-99-8
SnCl2
3260
[Cl-].[Cl-].[Sn++]
189.61
CHEBI:78067
98%
Tin(Ii) Chloride
Cl2Sn
MFCD00011241
AXZWODMDQAVCJE-UHFFFAOYSA-L
dichlorotin
24479
189.62g/mol
Reagent
Videos
Safety and Handling

Safety and Handling

P260-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338

H314-H317-H332-H335-H341-H361-H373-H410

missing translation for 'einecsNumber' : 231-868-0

missing translation for 'rtecsNumber' : XP8700000

SDS
Documents

Documents

Product Certifications