Learn More
Tin(II) fluoride, 99%, Thermo Scientific Chemicals
£14.90 - £170.85
Chemical Identifiers
CAS | 7783-47-3 |
---|---|
Molecular Formula | F2Sn |
Molecular Weight (g/mol) | 156.69 |
MDL Number | MFCD00042540 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10652345
|
Thermo Scientific Acros
308500050 |
5 g | Glass bottle | N/A | |||||
10450921
|
Thermo Scientific Acros
308501000 |
100 g | Plastic bottle |
£65.33
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10657494
|
Thermo Scientific Acros
308505000 |
500 g | Plastic bottle |
£170.85
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7783-47-3 | |
F2Sn | |
156.69 | |
MFCD00042540 |
Specifications
7783-47-3 | |
Powder, Crystals or Crystalline Powder | |
F2Sn | |
MFCD00042540 | |
Solubility in water: 300- 390g/L. Other solubilities: insoluble in ethanol,diethyl ether,chloroform | |
156.69 | |
Tin(II) fluoride |
White | |
99% | |
SnF2 | |
15, 8911 | |
156.69 | |
99% |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Causes skin irritation.
Causes serious eye damage.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 231-999-3
RUO – Research Use Only