Learn More
Tin(IV) chloride, 99%, anhydrous, Thermo Scientific Chemicals
£39.30 - £571.00
Chemical Identifiers
CAS | 7646-78-8 |
---|---|
Molecular Formula | Cl4Sn |
Molecular Weight (g/mol) | 260.52 |
InChI Key | HPGGPRDJHPYFRM-UHFFFAOYSA-J |
Synonym | tin tetrachloride, tin iv chloride, stannane, tetrachloro, tin chloride, stannic chloride, tetrachlorotin, tin perchloride, tintetrachloride, tin iv tetrachloride, sncl4 |
PubChem CID | 24287 |
IUPAC Name | tetrachlorostannane |
SMILES | Cl[Sn](Cl)(Cl)Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10472391
|
Thermo Scientific Acros
197510100 |
10 mL | Glass bottle |
£39.30
10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10482391
|
Thermo Scientific Acros
197511000 |
100 mL | Glass bottle |
£46.00
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10684702
|
Thermo Scientific Acros
197510010 |
1 L | Glass bottle |
£264.00
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11944181
|
Thermo Scientific Acros
197510025 |
2.5 L | Glass bottle |
£571.00
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acidChemical Identifiers
7646-78-8 | |
260.52 | |
tin tetrachloride, tin iv chloride, stannane, tetrachloro, tin chloride, stannic chloride, tetrachlorotin, tin perchloride, tintetrachloride, tin iv tetrachloride, sncl4 | |
tetrachlorostannane |
Cl4Sn | |
HPGGPRDJHPYFRM-UHFFFAOYSA-J | |
24287 | |
Cl[Sn](Cl)(Cl)Cl |
Specifications
114.0°C | |
7646-78-8 | |
Solution | |
Cl4Sn | |
01,1111; 03,269; 05,627; 06,553; 07,342; 09,436; 11,522; 12,486; 13,14; 14,304; 15,311; 17,335 | |
tin tetrachloride, tin iv chloride, stannane, tetrachloro, tin chloride, stannic chloride, tetrachlorotin, tin perchloride, tintetrachloride, tin iv tetrachloride, sncl4 | |
HPGGPRDJHPYFRM-UHFFFAOYSA-J | |
tetrachlorostannane | |
24287 | |
99% | |
2.2200g/mL |
-33.0°C | |
Brown-Red | |
98.5 to 101.5% (Argentometry) | |
SnCl4 | |
15,8901 | |
Solubility in water: reacts. Other solubilities: soluble in carbon disulfide, alcohol, carbon, tetrachloride, benzene, toluene, acetone, kerosene, and gasoline | |
Cl[Sn](Cl)(Cl)Cl | |
260.52 | |
260.52 | |
2.22 | |
Tin(IV) chloride |
Safety and Handling
GHS H Statement
Harmful to aquatic life with long lasting effects.
Causes severe skin burns and eye damage.
May cause respiratory irritation.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 231-588-9
Recommended Storage : Product may develop some turbidity or precipitate
- Tin(IV) chloride, anhydrous
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
Hazard Statement
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
- H412-Harmful to aquatic life with long lasting effects.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
RUO – Research Use Only