missing translation for 'onlineSavingsMsg'
Learn More

Tin(IV) chloride bis(2,4-pentanedionate), 95%, Thermo Scientific Chemicals

£97.08 - £356.00

Chemical Identifiers

CAS 16919-46-3
Molecular Formula C10H14Cl2O4Sn
Molecular Weight (g/mol) 387.83
MDL Number MFCD00015316
InChI Key DTMARTPDQMQTRX-VGKOASNMSA-J
Synonym Bis(acetylacetonato)dichlorotin(IV); Tin(IV) bis(acetylacetonate) chloride
SMILES [Cl-].[Cl-].[Sn+4].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11367298
View Documents
Thermo Scientific Alfa Aesar
030532.09
10 g
£97.08
10g
Please to purchase this item. Need a web account? Register with us today!
11377298
View Documents
Thermo Scientific Alfa Aesar
030532.18
50 g
£356.00
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

It is used as a raw material in synthesis. It is also used as NMR shift reagents and as catalysts for organic synthesis, and precursors to industrial hydroformylation catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

16919-46-3
387.83
DTMARTPDQMQTRX-VGKOASNMSA-J
[Cl-].[Cl-].[Sn+4].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
C10H14Cl2O4Sn
MFCD00015316
Bis(acetylacetonato)dichlorotin(IV); Tin(IV) bis(acetylacetonate) chloride

Specifications

199°C
Crystalline
MFCD00015316
Bis(acetylacetonato)dichlorotin(IV); Tin(IV) bis(acetylacetonate) chloride
DTMARTPDQMQTRX-VGKOASNMSA-J
tin(4+) bis((2Z)-4-oxopent-2-en-2-olate) dichloride
387.83
Moisture sensitive
16919-46-3
C10H14Cl2O4Sn
UN3146
Soluble in organic solvents.
[Cl-].[Cl-].[Sn+4].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
387.83
95%
Tin(IV) chloride bis(2,4-pentanedionate)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, SOLID, N.O.S.

missing translation for 'einecsNumber' : 240-972-5

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only