Learn More
Tin(IV) chloride hydrate, 98%, Thermo Scientific Chemicals
£26.93 - £92.76
Chemical Identifiers
CAS | 7646-78-8 |
---|---|
Molecular Formula | Cl4Sn |
Molecular Weight (g/mol) | 260.51 |
MDL Number | MFCD00149864 |
InChI Key | HPGGPRDJHPYFRM-UHFFFAOYSA-J |
Synonym | tin tetrachloride, tin iv chloride, stannane, tetrachloro, tin chloride, stannic chloride, tetrachlorotin, tin perchloride, tintetrachloride, tin iv tetrachloride, sncl4 |
PubChem CID | 24287 |
IUPAC Name | tetrachlorostannane |
SMILES | Cl[Sn](Cl)(Cl)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11337307
|
Thermo Scientific Alfa Aesar
011570.22 |
100 g |
£26.93
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11347307
|
Thermo Scientific Alfa Aesar
011570.36 |
500 g |
£92.76
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Tin(IV) chloride hydrate is used in the formation of a xerogel via a cyanogel, a gel that has potential use in solid-state gas sensors. It is involved in the preparation of Indium tin oxide thin films by chemical vapor deposition method.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7646-78-8 | |
260.51 | |
HPGGPRDJHPYFRM-UHFFFAOYSA-J | |
24287 | |
Cl[Sn](Cl)(Cl)Cl |
Cl4Sn | |
MFCD00149864 | |
tin tetrachloride, tin iv chloride, stannane, tetrachloro, tin chloride, stannic chloride, tetrachlorotin, tin perchloride, tintetrachloride, tin iv tetrachloride, sncl4 | |
tetrachlorostannane |
Specifications
56°C | |
Lump | |
Cl4Sn | |
tin tetrachloride, tin iv chloride, stannane, tetrachloro, tin chloride, stannic chloride, tetrachlorotin, tin perchloride, tintetrachloride, tin iv tetrachloride, sncl4 | |
HPGGPRDJHPYFRM-UHFFFAOYSA-J | |
tetrachlorostannane | |
24287 | |
Tin(IV) chloride hydrate |
7646-78-8 | |
98% | |
MFCD00149864 | |
Soluble in water and alcohol. | |
Cl[Sn](Cl)(Cl)Cl | |
260.51 | |
260.50 (Anhydrous) |
Safety and Handling
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: II
missing translation for 'einecsNumber' : 231-588-9
missing translation for 'rtecsNumber' : XP8750000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only