missing translation for 'onlineSavingsMsg'
Learn More

Cytiva TiO2 Mag Sepharose™

Designed for enrichment of phosphopeptides from tryptic digested protein samples. Cytiva TiO2 Mag Sepharose magnetic beads are based on titanium dioxide chromatography and are designed to simplify capture and enrichment of phosphopeptides.

£37.02 - £103.00

Specifications

Bead Type TiO2 Mag Sepharose
Matrix Paramagnetic spherical, highly cross linked agarose particles
Particle Size 37 to 100 μm
Storage Buffer 20% ethanol storage solution
Content And Storage 4°C to 30°C (Use at room temperature)
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10259734
View Documents Promotion Details
Cytiva
28-9440-10
500 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11590884
View Documents Promotion Details
Cytiva
28-9513-77
4 x 500 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

TiO2 Mag Sepharose has high affinity for phosphopeptides and is an efficient method for enrichment of phosphopeptides from complex samples. The magnetic bead format improves ease-of-use in sample preparation and has excellent properties for small-scale experiments. The high density beads allows rapid capture by magnetic devices with high visibility to ensure reliable collection of all bound target peptides.

  • Magnetic beads for selective capture and enrichment of phosphopeptides
  • Easy to use - visible and dense magnetic beads
  • Simple protocol with elution conditions optimized for MS analysis
  • Parallel sample handling when used with MagRack 6 separation tool - process up to six samples in less than one hour

 

Specifications

Specifications

TiO2 Mag Sepharose
37 to 100 μm
4°C to 30°C (Use at room temperature)
TiO2
Paramagnetic spherical, highly cross linked agarose particles
20% ethanol storage solution
Used for selective capture and enrichment of Phosphopeptides, for biomarker research
Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

SDS
Documents

Documents

Product Certifications