Learn More
Epredia™ Tissue-Loc™ HistoScreen™ Cassettes, Blue
Handle small biopsies and fragile specimens with Epredia™ Tissue-Loc™ HistoScreen™ Cassettes, the ultimate cassettes for specimen security.
Brand: Epredia™ C-1000-WH
This item is not returnable.
View return policy
Description
- Saves time, offers the greatest specimen security, and provides ease of use
- Provides an inner chamber lined with a “see-through" mesh for easy identification of specimens
- Eliminates the need for messy sponges or lens paper
- Ensures tissues are safely processed as cassettes will not pop open during processing
Specifications
Mesh Pore Insert | |
White | |
HistoScreen™ Cassette |