missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ TissueLoc™ Tissue Cassettes, White
Ensure specimen safety during processing and easy removal of cover while embedding with the hinge design of Epredia™ TissueLoc™ Tissue Cassettes.
Brand: Epredia™ 58960
This item is not returnable.
View return policy
Description
- Three-sided writing surface ensures positive identification from grossing to filing
- Solvent- and decalcifier-resistant and compatible with microwave techniques
- Eleven colors allow for easy specimen coding
Specifications
Slotted Pore | |
Hinged | |
Peach | |
Tissue Cassette |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction