missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ TissueLoc™ Tissue Cassettes, White

Ensure specimen safety during processing and easy removal of cover while embedding with the hinge design of Epredia™ TissueLoc™ Tissue Cassettes.

Brand:  Epredia™ 58955

 View more versions of this product

Product Code. 12697796

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Three-sided writing surface ensures positive identification from grossing to filing
  • Solvent- and decalcifier-resistant and compatible with microwave techniques
  • Eleven colors allow for easy specimen coding
Specifications

Specifications

Slotted Pore
Hinged
Blue
Tissue Cassette
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.