missing translation for 'onlineSavingsMsg'
Learn More
Titanium boride, 99.5% (metals basis), Thermo Scientific Chemicals Product Code.: 11415330

Titanium boride, 99.5% (metals basis), Thermo Scientific Chemicals

Product Code. 11415330
250 g
Click to view available options
Quantity:
250 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 11415330

Brand: Thermo Scientific Alfa Aesar 044558.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Used as a cathode material in aluminum smelting and can be shaped by electrical discharge machining. Specialized applications in such areas as impact resistant armor, cutting tools, crucibles, neutron absorbers and wear resistant coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12045-63-5
Molecular Formula B2Ti
Molecular Weight (g/mol) 69.487
MDL Number MFCD00049595
InChI Key QYEXBYZXHDUPRC-UHFFFAOYSA-N
Synonym titanium boride,
PubChem CID 131664302
IUPAC Name boron;titanium
SMILES [B].[B].[Ti]

Specifications

Melting Point 3225°C
Physical Form Powder
Quantity 250 g
Assay Percent Range 99.5% (metals basis)
Solubility Information Insoluble in water.
Formula Weight 69.5
Density 4.38
Chemical Name or Material Titanium boride

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberN07I020 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number044558.09,044558.18,044558.30
Lot NumberH30Q11 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number044558.09,044558.18,044558.30
Lot NumberU05H050 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number044558.09,044558.18,044558.30
Lot NumberK23U038 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number044558.09,044558.18,044558.30
Lot NumberL03U043 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number044558.09,044558.18,044558.30

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Titanium boride, 99.5% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.