Learn More
Titanium boride, 99.5% (metals basis), Thermo Scientific Chemicals
£174.87
Chemical Identifiers
CAS | 12045-63-5 |
---|---|
Molecular Formula | B2Ti |
Molecular Weight (g/mol) | 69.487 |
MDL Number | MFCD00049595 |
InChI Key | QYEXBYZXHDUPRC-UHFFFAOYSA-N |
Synonym | titanium boride, |
PubChem CID | 131664302 |
IUPAC Name | boron;titanium |
SMILES | [B].[B].[Ti] |
Description
Used as a cathode material in aluminum smelting and can be shaped by electrical discharge machining. Specialized applications in such areas as impact resistant armor, cutting tools, crucibles, neutron absorbers and wear resistant coatings.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12045-63-5 | |
69.487 | |
QYEXBYZXHDUPRC-UHFFFAOYSA-N | |
131664302 | |
[B].[B].[Ti] |
B2Ti | |
MFCD00049595 | |
titanium boride, | |
boron;titanium |
Specifications
3225°C | |
Powder | |
B2Ti | |
titanium boride, | |
QYEXBYZXHDUPRC-UHFFFAOYSA-N | |
boron;titanium | |
131664302 | |
4.38 |
12045-63-5 | |
99.5% (metals basis) | |
MFCD00049595 | |
Insoluble in water. | |
[B].[B].[Ti] | |
69.487 | |
69.5 | |
Titanium boride |
Safety and Handling
missing translation for 'einecsNumber' : 234-961-4
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only