Learn More
Titanium(II) hydride, Ti 94.0% min., Thermo Scientific Chemicals
£22.61 - £298.49
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11359399
|
Thermo Scientific Alfa Aesar
089183.06 |
5 g |
£22.61
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11369399
|
Thermo Scientific Alfa Aesar
089183.18 |
50 g |
£81.91
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11379399
|
Thermo Scientific Alfa Aesar
089183.30 |
250 g |
£298.49
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
For preparation of titanium compounds and coatings, in formation of glass and ceramic seals, as a catalyst, for thermal storage, as a source for pure H{2}Titanium(II) hydride is widely utilized as an additive in the manufacturing of Alnico sintered magnets and in sintered powdered metals. It is used in the production of metal foam, powdered titanium metal and in pyrotechnics. It is involved in various applications such as ceramics, pyrotechnics and sports equipment. It is also used as a laboratory reagent and as a blowing agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7704-98-5 | |
H2Ti | |
MFCD00151301 |
Specifications
450°C (decomposition) | |
Powder | |
H2Ti | |
Soluble in water. | |
Air and moisture sensitive | |
Titanium(II) Hydride |
7704-98-5 | |
99%, metal basis | |
MFCD00151301 | |
49.92 | |
3.9 |
Seguridad y manipulación
Take precautionary measures against static discharges.
P210-P240-P241-P280-P370+P378q
H228-H500
missing translation for 'dotInformation' : DOT Class: 4.1; Packaging Group: II
missing translation for 'einecsNumber' : 231-726-8
missing translation for 'rtecsNumber' : XR2130000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only