missing translation for 'onlineSavingsMsg'
Learn More
Titanium(II) oxide, 99.5% (metals basis), Thermo Scientific Chemicals Product Code.: 11334889

Titanium(II) oxide, 99.5% (metals basis), Thermo Scientific Chemicals

Product Code. 11334889
5 g
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11334889

Brand: Thermo Scientific Alfa Aesar 077126.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Titanium(II) oxide associated with vanadium oxide forms a transition-metal-oxide monolayer catalyst, which is used in the selective oxidation and ammoxidation of alkylaromatics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12137-20-1
Molecular Formula OTi
Molecular Weight (g/mol) 63.87
MDL Number MFCD00036279
InChI Key OGIDPMRJRNCKJF-UHFFFAOYSA-N
Synonym titanium monoxide, titanium ii oxide, titanium oxide tio, oxido titanium, titanium ii oxide pellets, titanium oxide, 99.9 %, titanium ii oxide, optical grade, titanium ii oxide trace metals basis 5g, titanium ii oxide, coating quality, granular, titanium ii oxide, pieces trace metals basisShow MoreShow Less
PubChem CID 61685
IUPAC Name oxotitanium
SMILES O=[Ti]

Specifications

Boiling Point >3000°C
Melting Point 1700°C
Physical Form Powder
Quantity 5 g
Assay Percent Range 99.5%, metal basis
Solubility Information Soluble in concentrated sulfuric acid,concentrated hydrofluoric acid and alkali. Insoluble in water,diluted sulfuric acid,hydrochloric acid,nitric acid and organic solvents.
Formula Weight 63.9
Density 4.95
Chemical Name or Material Titanium(II) Oxide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberM29K041 Certificate TypeCertificate of Analysis Date02/02/2024 Catalog Number077126.22,077126.06,077126.14,077126.36
Lot NumberS26J023 Certificate TypeCertificate of Analysis Date04/07/2023 Catalog Number077126.22,077126,077126.06,077126.14,077126.36
Lot NumberG05K22 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number077126.22,077126.06,077126.14,077126.36
Lot NumberB13S002 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number077126.22,077126.06,077126.14,077126.36
Lot NumberQ24B033 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number077126.22,077126.06,077126.14,077126.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Titanium(II) oxide, 99.5% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.