Learn More
Titanium(III) chloride-Aluminum(III) chloride, TiCl{3} 76.0-78.5%, Thermo Scientific Chemicals
Used as a catalyst in the production of polypropylene.
£109.55 - £442.20
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11304257
|
Thermo Scientific Alfa Aesar
036729.22 |
100 g |
£109.55
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11314257
|
Thermo Scientific Alfa Aesar
036729.36 |
500 g |
£442.20
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Titanium(III) chloride-Aluminum(III) chloride, TiCl{3} 76.0-78.5% is used as a catalyst in the production of polypropylene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12003-13-3 | |
MFCD00145412 |
Specifications
12003-13-3 | |
76.0 to 78.5% | |
MFCD00145412 | |
Insoluble in water. | |
Air and moisture sensitive |
Powder | |
(TiCl3)3:AlCl3 | |
Titanium aluminum chloride | |
596.15 | |
Titanium(III) Chloride-Aluminum(III) Chloride |
Safety and Handling
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Never add water to this product. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: II
missing translation for 'einecsNumber' : 234-421-8
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Argon
- Titanium(III) chloride-Aluminum(III) chloride
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501-Dispose of contents/container to .
RUO – Research Use Only