Learn More
Titanium(III) fluoride, Thermo Scientific Chemicals
Description
It is used as a laboratory chemicals & for manufacture of chemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Physical Form | Powder |
Quantity | 10 g |
UN Number | UN3260 |
Solubility Information | Sparingly soluble. It hydrolyses in water. |
Formula Weight | 104.9 |
Odor | Pungent, makes eyes water |
Sensitivity | Air and moisture sensitive |
Density | 3.400 g/mL |
Chemical Name or Material | Titanium(III) fluoride |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
Q19K028 | Certificate of Analysis | 11/06/2024 | 013079.09,013079.18 |
B14T032 | Certificate of Analysis | 07/12/2021 | 013079.09,013079.18 |
P03D009 | Certificate of Analysis | 07/12/2021 | 013079.09,013079.18 |
B07N21 | Certificate of Analysis | 07/12/2021 | 013079.09,013079.18 |
A08Z024 | Certificate of Analysis | 07/12/2021 | 013079.09,013079.18 |
Safety and Handling

- Titanium(III) fluoride
- Danger
- Skin corrosion/irritation Category 1 B
- H314-Causes severe skin burns and eye damage.
- EUH014-Reacts violently with water.
- EUH032-Contact with acids liberates very toxic gas.
- P233-Keep container tightly closed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P402+P404-Store in a dry place. Store in a closed container.
- MIXTURE LIST-Contains : Titanium fluoride
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.