missing translation for 'onlineSavingsMsg'
Learn More
Titanium(IV) ethoxide, 33-35% TiO2, Thermo Scientific Chemicals Product Code.: 11934151

Titanium(IV) ethoxide, 33-35% TiO2, Thermo Scientific Chemicals

Product Code. 11934151
1 kg, Glass bottle
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 11934151

Brand: Thermo Scientific Acros 223760010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3087-36-3
Molecular Formula C8H20O4Ti
Molecular Weight (g/mol) 228.15
InChI Key JMXKSZRRTHPKDL-UHFFFAOYSA-N
Synonym titanium iv ethoxide, ethyl titanate, titanium tetraethoxide, titanium ethoxide, tetraethyl titanate, titanium iv ethanolate, titanium 4+ tetraethanolate, titanium 4+ ethanolateShow MoreShow Less
PubChem CID 76524
IUPAC Name ethanolate;titanium(4+)
SMILES CC[O-].CC[O-].CC[O-].CC[O-].[Ti+4]

Specifications

Melting Point -40.0°C
Color Colorless to Yellow
Density 1.0900g/mL
Boiling Point 150.0°C to 152.0°C (10.0 mmHg)
Flash Point 33°C
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula Ti(OC2H5)4
Refractive Index 1.5050 to 1.5070
Quantity 1 kg
Beilstein 01,III,1338
Specific Gravity 1.09
Solubility Information Solubility in water: hydrolysis.
Viscosity 60-80 mPa.s (20°C)
Formula Weight 228.15
Percent Purity 33-35% TiO2
Physical Form Liquid
Chemical Name or Material Titanium(IV) ethoxide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0462951 Certificate TypeCertificate of Analysis Date18/06/2024 Catalog Number223760010,223762500,223760500
Lot NumberA0460922 Certificate TypeCertificate of Analysis Date13/03/2024 Catalog Number223760010,223762500,223760500
Lot NumberA0449201 Certificate TypeCertificate of Analysis Date04/01/2024 Catalog Number223760010,223762500,223760500
Lot NumberA0443751 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number223760010,223762500,223760500
Lot NumberA0389686 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number223760010,223762500,223760500
compliance-icons
Product Identifier
  • Titanium(IV) ethoxide
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Titanium(IV) ethoxide, 33-35% TiO2, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.