Learn More
Titanium(IV) methoxide, 95%, Thermo Scientific Chemicals
£76.40 - £903.00
Chemical Identifiers
CAS | 992-92-7 |
---|---|
Molecular Formula | Ti(OCH3)4·(CH3OH)x, x<1 |
MDL Number | MFCD00014883 |
Synonym | tetramethyl titanate, titanium tetramethoxide, titanium iv methoxide, methanolate; titanium 4+, titanium tetra methanolate, methanol, titanium 4+ salt, titanium methoxide, methanol, titanium 4+ salt 4:1, titanium 4+ tetrakis methoxide, titanium 4+ ion tetrakis methoxide |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11334899
|
Thermo Scientific Alfa Aesar
077137.14 |
25 g |
£76.40
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11344899
|
Thermo Scientific Alfa Aesar
077137.22 |
100 g |
£221.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11354899
|
Thermo Scientific Alfa Aesar
077137.36 |
500 g |
£903.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Titanium(IV) methoxide is used for the preparation of polyoxotitanates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
992-92-7 | |
Ti(OCH3)4·(CH3OH)x, x<1 | |
MFCD00014883 | |
tetramethyl titanate, titanium tetramethoxide, titanium iv methoxide, methanolate; titanium 4+, titanium tetra methanolate, methanol, titanium 4+ salt, titanium methoxide, methanol, titanium 4+ salt 4:1, titanium 4+ tetrakis methoxide, titanium 4+ ion tetrakis methoxide |
Specifications
>243°C (52mmHg) | |
992-92-7 | |
Ti(OCH3)4·(CH3OH)x, x<1 | |
UN3181 | |
tetramethyl titanate, titanium tetramethoxide, titanium iv methoxide, methanolate; titanium 4+, titanium tetra methanolate, methanol, titanium 4+ salt, titanium methoxide, methanol, titanium 4+ salt 4:1, titanium 4+ tetrakis methoxide, titanium 4+ ion tetrakis methoxide | |
95% | |
Titanium (IV) methoxide |
200°C to 210°C | |
Granular/Lump | |
MFCD00014883 | |
48°C (118°F) | |
Slightly soluble in organic solvents. | |
Moisture sensitive |
Safety and Handling
GHS H Statement
H228-H302-H312-H332-H319
Flammable solid.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.
P210-P240-P241-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P370+P378q-P501c
H228-H302+H312-H315-H318-H335
missing translation for 'dotInformation' : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.
missing translation for 'einecsNumber' : 213-596-4
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only