missing translation for 'onlineSavingsMsg'
Learn More
Titanium(IV) oxide, 98+%, anatase powder, Thermo Scientific Chemicals Product Code.: 10122392

Titanium(IV) oxide, 98+%, anatase powder, Thermo Scientific Chemicals

Product Code. 10122392
1 kg, Plastic bottle
Click to view available options
Quantity:
100 g
1 kg
Packaging:
Glass bottle
Plastic bottle
This item is not returnable. View return policy

Product Code. 10122392

Brand: Thermo Scientific Acros 213580010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13463-67-7
Molecular Formula O2Ti
Molecular Weight (g/mol) 79.87
MDL Number MFCD00011269 MFCD00210650
InChI Key GWEVSGVZZGPLCZ-UHFFFAOYSA-N
Synonym titanium dioxide, titania, titanium iv oxide, rutile, anatase, brookite, titanium white, flamenco, hombitan, titafrance
PubChem CID 26042
ChEBI CHEBI:32234
IUPAC Name dioxotitanium
SMILES O=[Ti]=O

Specifications

Boiling Point 2900.0°C
Melting Point 1855.0°C
Color White
pH 7.3 to 8.3 (aq. suspension)
Physical Form Powder
Quantity 1 kg
Assay Percent Range 98% min
Linear Formula TiO2
Merck Index 15, 9628
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in hot concentrated h2so4 and hf
Formula Weight 79.88
Percent Purity ≥98%
Packaging Plastic bottle
Specific Gravity 4.23
Density 4.2300g/mL
Chemical Name or Material Titanium(IV) oxide, 99%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0443806 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number213581000,213580010
Lot NumberA0402785 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number213581000,213580010
Lot NumberA0393899 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number213581000,213580010
Lot NumberA0402798 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number213581000,213580010
Lot NumberA0415063 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number213581000,213580010
Product Identifier
  • Titanium(IV) oxide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Titanium(IV) oxide, 98+%, anatase powder, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.