missing translation for 'onlineSavingsMsg'
Learn More
Titanium(IV) oxide, anatase, 99.6% (metals basis), Thermo Scientific Chemicals Product Code.: 11343238

Titanium(IV) oxide, anatase, 99.6% (metals basis), Thermo Scientific Chemicals

Product Code. 11343238
25 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11343238

Brand: Thermo Scientific Alfa Aesar 036199.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Titanium(IV) oxide is used as UV-resistant cloth, skin protecting cream, face washing milk, cream milk, powder make-up. It is also used as photocatalyst, self-cleaning glass, self-cleaning ceramics, antibacterial material, air purification, sewage handling, chemical industry. It is widely utilized in cosmetics, sunscreen cream, natural white moisture protection cream, beauty and whitening cream, dawn and night cream, moistening refresher, vanishing cream, chemical fiber, plastics, printing ink coating. It is likewise utilized for improving the impressionability and opacity of the paper and used for producing titanium, ferrotitanium alloy, carbide alloy etc. in the metallurgical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1317-70-0
Molecular Formula O2Ti
Molecular Weight (g/mol) 79.87
MDL Number MFCD00011269,MFCD00210650
InChI Key GWEVSGVZZGPLCZ-UHFFFAOYSA-N
Synonym titanium dioxide, titania, titanium iv oxide, rutile, anatase, brookite, titanium white, flamenco, hombitan, titafrance
PubChem CID 26042
ChEBI CHEBI:32234
IUPAC Name dioxotitanium
SMILES O=[Ti]=O

Specifications

Physical Form Powder
Quantity 25 g
Linear Formula TiO2
Merck Index 14,9472
Solubility Information Insoluble in water,dilute acids,organic acids,and dilute alkalis.
Formula Weight 79.9
Percent Purity 99.6%
Odor Odorless
Density 3.9 g/mL
Assay (metals basis)
Chemical Name or Material Titanium(IV) oxide, Anatase
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberM27L021 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number036199.22,036199.14,036199.36
Lot NumberM24L027 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog Number036199.22,036199.14,036199.36
Lot NumberU05K056 Certificate TypeCertificate of Analysis Date27/09/2024 Catalog Number036199.22,036199.14,036199.36
Lot NumberU14K011 Certificate TypeCertificate of Analysis Date06/09/2024 Catalog Number036199.22,036199.14,036199.36
Lot NumberY28J028 Certificate TypeCertificate of Analysis Date05/12/2023 Catalog Number036199.22,036199.14,036199.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Titanium(IV) oxide, anatase, 99.6% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.