missing translation for 'onlineSavingsMsg'
Learn More

Titanium(IV) oxide, NanoArc™, anatase, nanopowder, 99.9% (metals basis), Thermo Scientific Chemicals

£52.00 - £422.00

Chemical Identifiers

CAS 1317-70-0
Molecular Formula O2Ti
Molecular Weight (g/mol) 79.87
MDL Number MFCD00011269,MFCD00210650
InChI Key GWEVSGVZZGPLCZ-UHFFFAOYSA-N
Synonym titanium dioxide, titania, titanium iv oxide, rutile, anatase, brookite, titanium white, flamenco, hombitan, titafrance
PubChem CID 26042
ChEBI CHEBI:32234
IUPAC Name dioxotitanium
SMILES O=[Ti]=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11379147
View Documents
Thermo Scientific Alfa Aesar
039953.14
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11389147
View Documents
Thermo Scientific Alfa Aesar
100 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11399147
View Documents
Thermo Scientific Alfa Aesar
039953.36
500 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

It is used as UV-resistant cloth, skin protecting cream, face washing milk, cream milk, powder make-up. It is also used as photocatalyst, self-cleaning glass, self-cleaning ceramics, antibacterial material, air purification, sewage handling, chemical industry. It is widely utilized in cosmetics, sunscreen cream, natural white moisture protection cream, beauty and whitening cream, dawn and night cream, moistening refresher, vanishing cream, chemical fiber, plastics, printing ink coating. It is likewise utilized for improving the impressionability and opacity of the paper and used for producing titanium, ferrotitanium alloy, carbide alloy etc. in the metallurgical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1317-70-0
79.87
GWEVSGVZZGPLCZ-UHFFFAOYSA-N
26042
dioxotitanium
O2Ti
MFCD00011269,MFCD00210650
titanium dioxide, titania, titanium iv oxide, rutile, anatase, brookite, titanium white, flamenco, hombitan, titafrance
CHEBI:32234
O=[Ti]=O

Specifications

1317-70-0
99.9% (metals basis)
MFCD00011269,MFCD00210650
Insoluble in water,dilute acids,organic acids,and dilute alkalis.
O=[Ti]=O
79.87
CHEBI:32234
3.9 g/mL
Powder
O2Ti
titanium dioxide, titania, titanium iv oxide, rutile, anatase, brookite, titanium white, flamenco, hombitan, titafrance
GWEVSGVZZGPLCZ-UHFFFAOYSA-N
dioxotitanium
26042
79.9
Titanium(IV) oxide
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 215-280-1

missing translation for 'rtecsNumber' : BV6550000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only