missing translation for 'onlineSavingsMsg'
Learn More
Titanium(IV) oxide, NanoArc™, anatase, nanopowder, 99.9% (metals basis), Thermo Scientific Chemicals Product Code.: 11399147

Titanium(IV) oxide, NanoArc™, anatase, nanopowder, 99.9% (metals basis), Thermo Scientific Chemicals

Product Code. 11399147
500 g
Click to view available options
Quantity:
25 g
100 g
500 g
Este artículo no se puede devolver. Vea la política de devoluciones

Product Code. 11399147

Brand: Thermo Scientific Alfa Aesar 039953.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

Este artículo no se puede devolver. Vea la política de devoluciones

It is used as UV-resistant cloth, skin protecting cream, face washing milk, cream milk, powder make-up. It is also used as photocatalyst, self-cleaning glass, self-cleaning ceramics, antibacterial material, air purification, sewage handling, chemical industry. It is widely utilized in cosmetics, sunscreen cream, natural white moisture protection cream, beauty and whitening cream, dawn and night cream, moistening refresher, vanishing cream, chemical fiber, plastics, printing ink coating. It is likewise utilized for improving the impressionability and opacity of the paper and used for producing titanium, ferrotitanium alloy, carbide alloy etc. in the metallurgical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 1317-70-0
Molecular Formula O2Ti
Molecular Weight (g/mol) 79.87
MDL Number MFCD00011269,MFCD00210650
InChI Key GWEVSGVZZGPLCZ-UHFFFAOYSA-N
Synonym titanium dioxide, titania, titanium iv oxide, rutile, anatase, brookite, titanium white, flamenco, hombitan, titafrance
PubChem CID 26042
ChEBI CHEBI:32234
IUPAC Name dioxotitanium
SMILES O=[Ti]=O

Especificaciones

Physical Form Powder
Quantity 500 g
Assay Percent Range 99.9% (metals basis)
Solubility Information Insoluble in water,dilute acids,organic acids,and dilute alkalis.
Formula Weight 79.9
Density 3.9 g/mL
Chemical Name or Material Titanium(IV) oxide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberJ22W050 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number039953.22,039953.14,039953.36
Lot NumberU12D005 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number039953.22,039953.14,039953.36
Lot NumberN10C008 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number039953.22,039953.14,039953.36
Lot NumberZ17E024 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number039953.22,039953.14,039953.36
Lot NumberY12A054 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number039953.22,039953.14,039953.36

RUO – Research Use Only

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto
Titanium(IV) oxide, NanoArc™, anatase, nanopowder, 99.9% (metals basis), Thermo Scientific Chemicals >

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado