Learn More
Titanium(IV) oxide, anatase, 99.6% (metals basis), Thermo Scientific Chemicals
Description
Titanium(IV) oxide is used as UV-resistant cloth, skin protecting cream, face washing milk, cream milk, powder make-up. It is also used as photocatalyst, self-cleaning glass, self-cleaning ceramics, antibacterial material, air purification, sewage handling, chemical industry. It is widely utilized in cosmetics, sunscreen cream, natural white moisture protection cream, beauty and whitening cream, dawn and night cream, moistening refresher, vanishing cream, chemical fiber, plastics, printing ink coating. It is likewise utilized for improving the impressionability and opacity of the paper and used for producing titanium, ferrotitanium alloy, carbide alloy etc. in the metallurgical industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Physical Form | Powder |
Quantity | 500 g |
Linear Formula | TiO2 |
Merck Index | 14,9472 |
Solubility Information | Insoluble in water,dilute acids,organic acids,and dilute alkalis. |
Formula Weight | 79.9 |
Percent Purity | 99.6% |
Odor | Odorless |
Density | 3.9 g/mL |
Assay | (metals basis) |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
M27L021 | Certificate of Analysis | 29/01/2025 | 036199.22,036199.14,036199.36 |
M24L027 | Certificate of Analysis | 27/01/2025 | 036199.22,036199.14,036199.36 |
U05K056 | Certificate of Analysis | 27/09/2024 | 036199.22,036199.14,036199.36 |
U14K011 | Certificate of Analysis | 06/09/2024 | 036199.22,036199.14,036199.36 |
Y28J028 | Certificate of Analysis | 05/12/2023 | 036199.22,036199.14,036199.36 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.