missing translation for 'onlineSavingsMsg'
Learn More
Titanium(IV) oxide, catalyst support, Thermo Scientific Chemicals Product Code.: 11310488

Titanium(IV) oxide, catalyst support, Thermo Scientific Chemicals

Product Code. 11310488
5 kg
Click to view available options
Quantity:
250 g
1 kg
5 kg
This item is not returnable. View return policy

Product Code. 11310488

Brand: Thermo Scientific Alfa Aesar 043828.A7

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1317-70-0
Molecular Formula O2Ti
Molecular Weight (g/mol) 79.87
MDL Number MFCD00011269,MFCD00210650
InChI Key GWEVSGVZZGPLCZ-UHFFFAOYSA-N
Synonym titanium dioxide, titania, titanium iv oxide, rutile, anatase, brookite, titanium white, flamenco, hombitan, titafrance
PubChem CID 26042
ChEBI CHEBI:32234
IUPAC Name dioxotitanium
SMILES O=[Ti]=O

Specifications

Physical Form Pellets
Quantity 5 kg
Formula Weight 79.9
Chemical Name or Material Titanium(IV) Oxide, Catalyst Support

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberM23J043 Certificate TypeCertificate of Analysis Date27/01/2023 Catalog Number043828.30,043828.A1,043828.A7
Lot NumberR01F016 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number043828.30,043828.A1,043828.A7
Lot NumberZ01H033 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number043828.30,043828.A1,043828.A7
Lot NumberY03H009 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number043828.30,043828.A1,043828.A7
Lot NumberN19F029 Certificate TypeCertificate of Analysis Date28/12/2021 Catalog Number043828.30,043828.A1,043828.A7

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Titanium(IV) oxide, catalyst support, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.