missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ TM2 TEAD inhibitor
Potent and reversible inhibitor of TEAD mediated Hippo/Yap signalling
£106.91 - £404.96
Specifications
Chemical Name or Material | 4-[3-(2-Cyclohexylethoxy)benzoyl]-N-phenyl-1-piperazinecarboxamide |
---|---|
CAS | 1008768-41-9 |
Target | YAP1 |
Molecular Formula | C26H33N3O3 |
Purity | 0.98 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18706075
|
Tocris Bioscience™
7917/10 |
10 mg |
£106.91
10mg |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18705925
|
Tocris Bioscience™
7917/50 |
50 mg |
£404.96
50mg |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
TM2 TEAD inhibitor is a potent and reversible TEA domain transcription factor inhibitor (IC50 = 38 nM and 156 nM for TEAD4 auto-palmitoylation and TEAD2 palmitoylation, respectively). It inhibits TEAD-YAP association and transcriptional activity. TM2 TEAD inhibitor exhibits strong antiproliferation effects as a single agent or in combination with a MEK inhibitor in YAP-dependent cancer cells. Also inhibits YAP-dependent liver organoid growth ex vivo.Specifications
4-[3-(2-Cyclohexylethoxy)benzoyl]-N-phenyl-1-piperazinecarboxamide | |
YAP1 | |
0.98 |
1008768-41-9 | |
C26H33N3O3 |