Learn More
TNC/Tenascin-C Rabbit, anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27789-1-AP-20UL
Description
The TNC/Tenascin-C antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human TNC/Tenascin-C. This antibody recognizes human antigen. The TNC/Tenascin-C antibody has been validated for the following applications: WB,ELISA analysis.
Specifications
TNC/Tenascin-C | |
Polyclonal | |
Unconjugated | |
20 μL | |
3371 | |
Liquid |
Western Blot, ELISA | |
450 μg/mL | |
Rabbit | |
Primary | |
Human |
Product Suggestions
Customers who viewed this item also viewed