Learn More
α-Tocopherol phosphate, MedChemExpress
MedChemExpress α-Tocopherol phosphate is the compound demonstrating the highest vitamin E activity, which is available both in its natural form as RRR-alpha-tocopherol isolated from plant sources.
Brand: Medchem Express HY-16686
This item is not returnable.
View return policy
Description
α-Tocopherol phosphate is the compound demonstrating the highest vitamin E activity, which is available both in its natural form as RRR-alpha-tocopherol isolated from plant sources.Specifications
α-Tocopherol phosphate | |
White | |
C29H51O5P | |
alpha-Tocopherol phosphate TocP vitamin E phosphate | |
CC1=C(O[C@@](CCC[C@H](C)CCC[C@H](C)CCCC(C)C)(C)CC2)C2=C(C)C(OP(O)(O)=O)=C1C | |
510.69 | |
Research |
38976-17-9 | |
Research | |
10 mg | |
DMSO : ≥ 100 mg/mL (195.81 mM) ∣H2O : 0.67 mg/mL (1.31 mM; Need ultrasonic) | |
510.69 | |
98.0% | |
Solid |