missing translation for 'onlineSavingsMsg'
Learn More

α-Tocopherol phosphate, MedChemExpress

MedChemExpress α-Tocopherol phosphate is the compound demonstrating the highest vitamin E activity, which is available both in its natural form as RRR-alpha-tocopherol isolated from plant sources.

£62.92 - £102.85

Specifications

Chemical Name or Material α-Tocopherol phosphate
CAS 38976-17-9
Color White
Purity Grade Notes Research
Molecular Formula C29H51O5P
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16459667
Encompass_Preferred
View Documents
Medchem Express
HY-16686
5 mg
£62.92
5mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16457128
Encompass_Preferred
View Documents
Medchem Express
HY-16686
10 mg
£102.85
10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

α-Tocopherol phosphate is the compound demonstrating the highest vitamin E activity, which is available both in its natural form as RRR-alpha-tocopherol isolated from plant sources.
Specifications

Specifications

α-Tocopherol phosphate
White
C29H51O5P
DMSO : ≥ 100 mg/mL (195.81 mM) ∣H2O : 0.67 mg/mL (1.31 mM; Need ultrasonic)
510.69
98.0%
Solid
38976-17-9
Research
alpha-Tocopherol phosphate TocP vitamin E phosphate
CC1=C(O[C@@](CCC[C@H](C)CCC[C@H](C)CCCC(C)C)(C)CC2)C2=C(C)C(OP(O)(O)=O)=C1C
510.69
Research
Videos
Safety and Handling

Safety and Handling

H315∣H319

missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

SDS
Documents

Documents

Product Certifications

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.