missing translation for 'onlineSavingsMsg'
Learn More
Toluene, 99.85%, for HPLC, Thermo Scientific Chemicals Product Code.: 10114730

Toluene, 99.85%, for HPLC, Thermo Scientific Chemicals

Product Code. 10114730
2.5 L, Glass Bottle
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Product Code. 10114730

Brand: Thermo Scientific Acros 268370025

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For HPLC
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-88-3
Molecular Formula C7H8
Molecular Weight (g/mol) 92.14
MDL Number MFCD00008512
InChI Key YXFVVABEGXRONW-UHFFFAOYSA-N
Synonym methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzeneShow MoreShow Less
PubChem CID 1140
ChEBI CHEBI:17578
IUPAC Name toluene
SMILES CC1=CC=CC=C1

Specifications

Absorbance (1 cm cell vs HPLC−grade water), 0.01 max. at 350nm, 0.02 max. at 340nm, 0.03 max. at 330nm, 0.05 max. at 310nm, 0.05 max. at 320nm, 0.1 max. at 300nm, 0.5 max. at 290nm, cut off at 286nm
Acidity 0.0015 meq/g max.
Density 0.8660g/mL
Quantity 2.5 L
Packaging Glass Bottle
Physical Form Liquid
Assay Percent Range 99.85%
Residue after Evaporation 0.0002% max.
Linear Formula C6H5CH3
Vapor Pressure Vapor Pressure: 29mbar at 20°C
Beilstein 05, 280
Water 0.03% max.
Merck Index 15, 9688
Formula Weight 92.14
Percent Purity ≥99.8%
Grade HPLC
Chemical Name or Material Toluene
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2533994 Certificate TypeCertificate of Analysis Date10/04/2025 Catalog Number268370025,268370010
Lot Number2404618 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number268370025,268370010
Lot Number2401187 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number268370025,268370010
Lot Number2411693 Certificate TypeCertificate of Analysis Date30/01/2025 Catalog Number268370025,268370010
Lot Number2416687 Certificate TypeCertificate of Analysis Date30/01/2025 Catalog Number268370025,268370010
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Toluene
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H361d-Suspected of damaging the unborn child.
  • H373-May cause damage to organs.
  • H336-May cause drowsiness or dizziness.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P331-Do NOT induce vomiting.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Toluene, 99.85%, for HPLC, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.