missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Toluene, CHROMASOLV™, for HPLC, 99.9%, Honeywell Riedel-de Haen
CHROMASOLV™, for HPLC, 99.9%
£25.97 - £241.00
Chemical Identifiers
CAS | 108-88-3 |
---|---|
Molecular Formula | C7H8 |
Molecular Weight (g/mol) | 92.14 |
MDL Number | MFCD00008512 |
InChI Key | YXFVVABEGXRONW-UHFFFAOYSA-N |
Synonym | methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene |
PubChem CID | 1140 |
ChEBI | CHEBI:17578 |
IUPAC Name | toluene |
SMILES | CC1=CC=CC=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15624610
|
Honeywell-Chromasolv
34866-100ML |
100 mL | Glass Bottle | ||||||
Currently Unavailable: We have no information when this product will be available
|
15634610
|
Honeywell-Chromasolv
34866-1L |
1 L | Glass Bottle |
£25.97
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15654610
|
Honeywell-Chromasolv
34866-2L |
2 L | Glass Bottle | ||||||
15644610
|
Honeywell-Chromasolv
34866-2.5L |
2.5 L | Glass Bottle |
£241.00
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15674610
|
Honeywell-Chromasolv
34866-6X1L |
6 x 1 L | Glass Bottle |
£209.35
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15664610
|
Honeywell-Chromasolv
34866-4X2.5L |
4 x 2.5 L | Glass Bottle |
£208.21
Pack of 4 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- For HPLC and organic synthesis
- High-purity, multipurpose solvents
- High lot-to-lot consistency
Chemical Identifiers
108-88-3 | |
92.14 | |
YXFVVABEGXRONW-UHFFFAOYSA-N | |
1140 | |
toluene |
C7H8 | |
MFCD00008512 | |
methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene | |
CHEBI:17578 | |
CC1=CC=CC=C1 |
Specifications
-93°C (literature) | |
108-88-3 | |
0.865 g/mL at 25 °C(lit.) | |
4°C | |
C6H5CH3 | |
UN1294 | |
methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene | |
CC1=CC=CC=C1 | |
92.14 | |
CHEBI:17578 | |
>99.9% | |
Toluene, CHROMASOLV™, for HPLC, 99.9% |
max. 1.00 at 286 nm, max. 0.01 at 400 nm, max. 0.01 at 350 nm, max. 0.10 at 300 nm, max. 0.50 at 288 nm | |
100.0000% | |
110.6°C | |
C7H8 | |
MFCD00008512 | |
635760 | |
YXFVVABEGXRONW-UHFFFAOYSA-N | |
toluene | |
1140 | |
92.14 | |
HPLC |
Safety and Handling
missing translation for 'shelfLife' : 1800 days from date of manufacture
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Toluene
missing translation for 'einecsNumber' : 203-625-9
Recommended Storage : Room Temp
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Toluene
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Aspiration toxicity Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- Reproductive toxicity Category 2
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H361d-Suspected of damaging the unborn child.
- H373-May cause damage to organs.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.