missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Toluene, Laboratory Reagent, ≥99.3%, Honeywell™
Laboratory Reagent,≥99.3%
£37.14 - £384.00
Chemical Identifiers
CAS | 108-88-3 |
---|---|
Molecular Formula | C7H8 |
Molecular Weight (g/mol) | 92.14 |
MDL Number | MFCD00008512 |
InChI Key | YXFVVABEGXRONW-UHFFFAOYSA-N |
Synonym | methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene |
PubChem CID | 1140 |
ChEBI | CHEBI:17578 |
IUPAC Name | toluene |
SMILES | CC1=CC=CC=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15681630
|
Honeywell Research Chemicals
179965-1L |
1 L | Glass Bottle |
£37.14
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15691630
|
Honeywell Research Chemicals
179965-2.5L |
2.5 L | Glass Bottle |
£81.21
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15621640
|
Honeywell Research Chemicals
179965-6X1L |
6 x 1 L | Glass Bottle |
£182.48
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15671630
|
Honeywell Research Chemicals
179965-10L |
10 L | Stainless Steel Can |
£262.26
10 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15601640
|
Honeywell Research Chemicals
179965-20L |
20 L | Stainless Steel Can | ||||||
Description
- ≥95% for general laboratory use.
Chemical Identifiers
108-88-3 | |
92.14 | |
YXFVVABEGXRONW-UHFFFAOYSA-N | |
1140 | |
toluene |
C7H8 | |
MFCD00008512 | |
methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene | |
CHEBI:17578 | |
CC1=CC=CC=C1 |
Specifications
-95°C | |
Colorless | |
110.6°C | |
4°C (39.2°F) | |
Liquid | |
C7H8 | |
MFCD00008512 | |
UN1294 | |
methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene | |
CC1=CC=CC=C1 | |
92.14 | |
CHEBI:17578 | |
≥99.3% | |
Toluene |
108-88-3 | |
0.865g/mL at 25°C | |
0.07 | |
997°F | |
n/D 1.496(lit.) | |
C6H5CH3 | |
22mmHg (20°C), 26mmHg (25°C) | |
635760 | |
YXFVVABEGXRONW-UHFFFAOYSA-N | |
toluene | |
1140 | |
92.14g/mol | |
Laboratory Reagent |
Safety and Handling
P210-P260-P280-P301 + P310-P370 + P378-P403 + P235
H225-H304-H315-H336-H361d-H373
missing translation for 'dotInformation' : class 3 - PG 2 - Toluene
missing translation for 'einecsNumber' : 203-625-9
missing translation for 'rtecsNumber' : XS5250000