missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TOPO™ XL-2 Complete PCR Cloning Kit, without competent cells
Complete PCR Cloning Kit provides all the necessary elements for efficient cloning of extra-long PCR products
Brand: Invitrogen™ K8040-10
214.50 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.
This item is not returnable.
View return policy
Description
- The TOPO™ XL-2 Complete PCR Cloning Kit provides all the necessary elements for efficient cloning of extra-long PCR products (up to 13 kb).
- The kit uses the linearized and topoisomerase 1-activated pCR™-XL-2-TOPO™ vector, which is compatible with the cloning of blunt-end PCR fragments. Amplification of long PCR fragments is enabled by Platinum™ SuperFi™ Green PCR Master Mix, which is included in the kit.
- Topoisomerase I activation of the vector allows PCR products to be ligated in just 5 minutes on your bench–top, resulting in high cloning efficiency (up to 90% positive recombinants with 10 kb fragments).
- ccdB gene for positive selection
- EcoR I site flanking the PCR product insertion site for easy excision of inserts
- Ampicillin- and kanamycin-resistance genes for your choice of selection in E. coli
- T7 promoter/priming site for in vitro RNA transcription and sequencing
- T7, T3, and M13 forward and reverse primer sites for sequencing
- TOPO XL-2 PCR Cloning Kits containing the pCR-XL-2-TOPO vector
- Platinum SuperFi Green PCR Master Mix—featuring a proofreading DNA polymerase (>100X fidelity compared to Taq) with high processivity and a density gradient to generate accurate, long PCR amplicons ready to load onto an agarose gel for gel extraction
- PureLink™ Quick Gel Extraction and PCR Purification Combo Kit—designed to purify DNA fragments from agarose gels or direct PCR purification using a silica-based spin cartridge in < 30 minutes
Specifications
Not Included | |
Zero Blunt TOPO | |
Kit | |
10 Reactions | |
Cloning Kit | |
10 reactions | |
pCR-XL-2-TOPO |
Ampicillin (AmpR), Kanamycin (KanR) | |
Box 1: Store at -5 to -30°C. • pCR XL-2-TOPO vector • Salt Solution • T3 Primer • T7 Primer • XL Control PCR Template • XL Control PCR Primers • Platinum SuperFi Green PCR Master Mix • SuperFi GC Enhancer • Nuclease-free water Box 2: Store at room temperature. • Gel Solubilization Buffer (L3) • Binding Buffer (B2) • Wash Buffer (W1) • Elution Buffer (E1) • PureLink Clean-up Spin Columns (in Wash Tubes) • PureLink Recovery/Elution Tubes |
|
PCR Cloning | |
TOPO™ | |
T3, T7 | |
Approved for shipment at Room Temperature or on Wet or Dry Ice |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction