missing translation for 'onlineSavingsMsg'
Learn More

Unity Lab Services™ Total Care Service Plan for Thermo Scientific Refrigerators & Freezers (Down to -40°C)

Total Care Service Plan provides support post factory warranty. Includes 2 day response time (within specified location and criteria), full parts and labor coverage, and one preventative maintenance visit.

Brand:  Unity Lab Services™ TLCLRF

Product Code. 17250444

  • £1034.00 / Each
Estimated Shipment: 02-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Continue comprehensive service support post factory warranty with faster response time, and proactive maintenance through annual preventative maintenance visit (or event)
  • 2 business day response time (may vary by location) for corrective maintenance
  • Includes Engineer labor, travel, and factory-certified replacement parts required for corrective maintenance
  • One scheduled Preventative Maintenance visit including a preventative maintenance kit (where applicable)
  • Priority unlimited access to OEM-trained Technical support experts who can diagnose, investigate, and remotely resolve issues or schedule on-site visits using the latest digital remote support tools
  • Access to on-line service knowledge base
  • Corrective Maintenance Repair Guarantee for the duration of product warranty or 1 year whichever is longer, as defined in the contract terms and conditions
  • Instrument Software/ Firmware updates included as needed to support a preventative maintenance or corrective maintenance visit
  • Upgrades to new versions of software are not included
  • 10% discount on additional services including additional preventative maintenance visits, calibration, and qualification services
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Support Post Factory Warranty
Full Parts & Labor Coverage, and One Preventative Maintenance Visit
2 Days (Within Specified Location and Criteria)
Total Care Service Plan
2 Day Response Time (Within Specified Location and Criteria), Full Parts & Labor Coverage, and One Preventative Maintenance Visit
Upgrades to New Software Versions
Refrigerators and Freezers (Down to -40°C)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.